2,2-Diethoxypropane ≥95.0%
Supplier: TCI America
Synonyms:
Acetone diethyl Acetal
D0459-10ML
D0459-250ML
TCD0459-010MLEA
79.26
USD
TCD0459-010ML
TCD0459-250ML
2,2-Diethoxypropane ≥95.0%
2,2-Diethoxypropane
CAS Number: 126-84-1
MDL Number: MFCD00009224
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 7
Specific Gravity (20/20): 0.83
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Acetone Diethyl Acetal
Formula:
C₇H₁₆O₂ Boiling Pt: 113 °C Density: 0.83 (20 °C) Flash Pt: 7 °C |
MDL Number:
MFCD00009224 CAS Number: 126-84-1 EINECS: 204-808-6 UN: 3271 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...