N,N-Dimethylglycine methyl ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0257-500G D0257-25G
TCD0257-500GEA 720 USD
TCD0257-500G TCD0257-025G
N,N-Dimethylglycine methyl ester ≥98.0% (by GC, titration analysis)
N,N-Dimethylglycine methyl ester

CAS Number: 7148-06-3
MDL Number: MFCD00059303
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C

Formula: C₅H₁₁NO₂
Boiling Pt: 135 °C
Density: 0.95 (20 °C)
MDL Number: MFCD00059303
CAS Number: 7148-06-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0288f5dfc4f3aa513ec9dbeeeacc1c6c891f6a9d&vid=0288f5dfc4f3aa513ec9dbeeeacc1c6c891f6a9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR