2-(Dibutylamino)ethanol ≥99.0%
Supplier: TCI America
Synonyms:
N,N-Dibutylethanolamine
D0219-500ML
D0219-25ML
TCD0219-500MLEA
50.31
USD
TCD0219-500ML
TCD0219-025ML
2-(Dibutylamino)ethanol ≥99.0%
2-(Dibutylamino)ethanol
CAS Number: 102-81-8
MDL Number: MFCD00014033
Molecular Formula: C10H23NO
Molecular Weight: 173.30
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Melting point (°C): -70
Flash Point (°C): 90
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: N,N-Dibutylethanolamine
Formula:
C₁₀H₂₃NO MW: 173.3 g/mol Boiling Pt: 226 °C Melting Pt: -70 °C Density: 0.86 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00014033 CAS Number: 102-81-8 EINECS: 203-057-1 UN: 2873 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...