meso-2,3-Dibromosuccinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0211-25G D0211-500G
TCD0211-025GEA 20 USD
TCD0211-025G TCD0211-500G
meso-2,3-Dibromosuccinic acid ≥98.0% (by titrimetric analysis)
meso-2,3-Dibromosuccinic acid

CAS Number: 608-36-6
MDL Number: MFCD00066439
Molecular Formula: C4H4Br2O4
Molecular Weight: 275.88
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 256

Formula: HOOC(BrCH)₂COOH
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 256 °C
MDL Number: MFCD00066439
CAS Number: 608-36-6
EINECS: 452-290-7
UN: 3261
ADR: 8,III
Merck Index: 13,03054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=221bc5da951114ac8b18164d299fac82f61fc4d4&vid=221bc5da951114ac8b18164d299fac82f61fc4d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR