1,4-Diacetylbenzene ≥99.0%
Supplier: TCI America
Synonyms:
4'-Acetylacetophenone
D0066-5G
D0066-25G
TCD0066-005GEA
65.57
USD
TCD0066-005G
TCD0066-025G
1,4-Diacetylbenzene ≥99.0%
1,4-Diacetylbenzene
CAS Number: 1009-61-6
MDL Number: MFCD00008750
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 113
Other Product Names / Synonyms: 4'-Acetylacetophenone
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Boiling Pt: 80 Melting Pt: 113 °C |
MDL Number:
MFCD00008750 CAS Number: 1009-61-6 EINECS: 213-769-4 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...