4-Chloro-2-fluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C2264-5G
C2264-25G
TCC2264-5GEA
79
USD
TCC2264-5G
TCC2264-25G
4-Chloro-2-fluorophenol ≥98.0% (by GC, titration analysis)
4-Chloro-2-fluorophenol
CAS Number: 348-62-9
MDL Number: MFCD00075030
Molecular Formula: C6H4ClFO
Molecular Weight: 146.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 104
Melting point (°C): 20
Flash Point (°C): 81
Specific Gravity (20/20): 1.40
Formula:
C₆H₄ClFO MW: 146.55 g/mol Boiling Pt: 104 °C Melting Pt: 20 °C Density: 1.40 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00075030 CAS Number: 348-62-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...