(R)-(+)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
(R)-(+)-2-Chloropropanoic acid
C1655-1G
C1655-5G
C1655-25G
TCC1655-001GEA
20
USD
TCC1655-001G
TCC1655-005G
TCC1655-25G
(R)-(+)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-2-Chloropropionic acid
CAS Number: 7474-05-7
MDL Number: MFCD00064204
Molecular Formula: C3H5ClO2
Molecular Weight: 108.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 107
Specific Gravity (20/20): 1.26
Specific rotation [a]20/D: 14 deg (neat)
Formula:
H₃CCHClCOOH MW: 108.52 g/mol Boiling Pt: 185 °C (1013 hPa) Melting Pt: –12 °C Density: 1.26 (20 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064204 CAS Number: 7474-05-7 UN: 2511 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...