2-Pyrimidinecarbonitrile ≥98.0%
Supplier: TCI America
Synonyms:
2-Cyanopyrimidine
C1518-25G
C1518-5G
C1518-1G
TCC1518-25GEA
237
USD
TCC1518-25G
TCC1518-5G
TCC1518-001G
2-Pyrimidinecarbonitrile ≥98.0%
2-Pyrimidinecarbonitrile
CAS Number: 14080-23-0
MDL Number: MFCD00160513
Molecular Formula: C5H3N3
Molecular Weight: 105.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41
Other Product Names / Synonyms: 2-Pyrimidinecarbonitrile
Formula:
C₅H₃N₃ MW: 105.1 g/mol Melting Pt: 41 °C |
MDL Number:
MFCD00160513 CAS Number: 14080-23-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...