Hexyl hexanoate ≥98.0%

Supplier: TCI America

Synonyms: Hexanoic acid hexyl ester

C1514-25ML C1514-500ML
TCC1514-025MLEA 39.69 USD
TCC1514-025ML TCC1514-500ML
Hexyl hexanoate ≥98.0%
Hexyl hexanoate

CAS Number: 6378-65-0
MDL Number: MFCD00053808
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 244
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Hexanoic Acid Hexyl Ester

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 244 °C
Melting Pt: –55 °C
Density: 0.86 (20 °C)
MDL Number: MFCD00053808
CAS Number: 6378-65-0
EINECS: 228-952-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ed312b075ccc496d9b9f23030d880926edf41f3&vid=0ed312b075ccc496d9b9f23030d880926edf41f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR