7-Chloroquinaldine ≥98.0%

Supplier: TCI America

Synonyms: 7-Chloro-2-methylquinoline

C1453-5G C1453-25G
TCC1453-005GEA 27 USD
TCC1453-005G TCC1453-025G
7-Chloroquinaldine ≥98.0%
7-Chloroquinaldine

CAS Number: 4965-33-7
MDL Number: MFCD00075436
Molecular Formula: C10H8ClN
Molecular Weight: 177.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 87
Melting point (°C): 76


Other Product Names / Synonyms: 7-Chloro-2-methylquinoline

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 87 °C
Melting Pt: 76 °C
MDL Number: MFCD00075436
CAS Number: 4965-33-7
EINECS: 440-600-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db5fe812120177afde25f25e236bb6fe1fee5a7&vid=5db5fe812120177afde25f25e236bb6fe1fee5a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR