4-(Chloromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1283-25G C1283-500G C1283-100G
TCC1283-025GEA 67 USD
TCC1283-025G TCC1283-500G TCC1283-100G
4-(Chloromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)
4-(Chloromethyl)benzoyl chloride

CAS Number: 876-08-4
MDL Number: MFCD00053224
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 93
Freezing point (°C): 27

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 1.3034 g/cm³ (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00053224
CAS Number: 876-08-4
EINECS: 212-881-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75d0ac0ad3dc1487cef0dd58d672c6f53d692726&vid=75d0ac0ad3dc1487cef0dd58d672c6f53d692726&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR