3-Chloro-1-butyne ≥98.0%

Supplier: TCI America

C1195-1G C1195-5G
TCC1195-001GEA 74 USD
TCC1195-001G TCC1195-5G
3-Chloro-1-butyne ≥98.0%
3-Chloro-1-butyne

CAS Number: 21020-24-6
MDL Number: MFCD00143109
Molecular Formula: C4H5Cl
Molecular Weight: 88.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Specific Gravity (20/20): 0.96

Formula: C₄H₅Cl
MW: 88.54 g/mol
Boiling Pt: 68 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00143109
CAS Number: 21020-24-6
EINECS: 244-151-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d76161abcf9cab064ea8cd27b7b40e1e6691a793&vid=d76161abcf9cab064ea8cd27b7b40e1e6691a793&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR