Allyl cyanoacetate ≥97.0%

Supplier: TCI America

Synonyms: Cyanoacetic acid Allyl Ester, Allyl Cyanoacetate, Allyl 2-cyanoacetate, Cyanoacetic acid allyl ester

C1039-500G C1039-25G
TCC1039-500GEA 344.95 USD
TCC1039-500G TCC1039-025G
Allyl cyanoacetate ≥97.0%
Allyl cyanoacetate

CAS Number: 13361-32-5
MDL Number: MFCD00013817
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 113
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cyanoacetic Acid Allyl Ester

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 101 °C
Melting Pt: –40 °C
Density: 1.07 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00013817
CAS Number: 13361-32-5
EINECS: 236-423-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d8e3551272b84bc05bac392a00a3bd2d1497e0b&vid=1d8e3551272b84bc05bac392a00a3bd2d1497e0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR