1-Cyclopropylethanol ≥98.0%

Supplier: TCI America

Synonyms: Cyclopropylmethylcarbinol

C0984-5ML C0984-25ML
TCC0984-005MLEA 42 USD
TCC0984-005ML TCC0984-025ML
1-Cyclopropylethanol ≥98.0%
1-Cyclopropylethanol

CAS Number: 765-42-4
MDL Number: MFCD00001300
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 31
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Cyclopropylmethylcarbinol

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 122 °C
Melting Pt: –31.4 °C
Density: 0.89 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00001300
CAS Number: 765-42-4
EINECS: 212-145-9
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5184fee856c8dace6af45d49df11d696f229e85e&vid=5184fee856c8dace6af45d49df11d696f229e85e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR