2-Cyclohexen-1-ol ≥95.0%
Supplier: TCI America
Synonyms:
Cyclohex-2-ene-1-ol, Cyclohex-2-enol
C0816-5ML
C0816-25ML
TCC0816-005MLEA
174
USD
TCC0816-005ML
TCC0816-25ML
2-Cyclohexen-1-ol ≥95.0%
2-Cyclohexen-1-ol
CAS Number: 822-67-3
MDL Number: MFCD00001570
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Flash Point (°C): 58
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 1,2,3,4-Tetrahydrophenol
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 166 °C Density: 0.99 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00001570 CAS Number: 822-67-3 EINECS: 212-501-3 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...