Cycloheptene ≥96.0%

Supplier: TCI America

C0688-5ML C0688-25ML
TCC0688-005MLEA 83 USD
TCC0688-005ML TCC0688-025ML
Cycloheptene ≥96.0%
Cycloheptene

CAS Number: 628-92-2
MDL Number: MFCD00004156
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 23
Specific Gravity (20/20): 0.83

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 115 °C
Melting Pt: -56 °C
Density: 0.83 (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07be40630d09be7cd15f8ee7a83420ff0b5db464&vid=07be40630d09be7cd15f8ee7a83420ff0b5db464&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR