3-Chloro-1-butene ≥98.0%

Supplier: TCI America

C0581-25G C0581-100G
TCC0581-025GEA 324 USD
TCC0581-025G TCC0581-100G
3-Chloro-1-butene ≥98.0%
3-Chloro-1-butene

CAS Number: 563-52-0
MDL Number: MFCD00000869
Molecular Formula: C4H7Cl
Molecular Weight: 90.55
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): -27
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 64 °C
Melting Pt: -50 °C
Density: 0.90 (20 °C)
Flash Pt: -27 °C
MDL Number: MFCD00000869
CAS Number: 563-52-0
EINECS: 209-252-8
UN: 1993
ADR: 3,II
Merck Index: 13,02150
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed18659dd1db720c8e8163873bf563fdab34e33&vid=98d00e277b95fdb91509e6cd13d0dfc1c72d4238&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR