Cyclooctene ≥95.0%

Supplier: TCI America

C0506-25ML C0506-500ML
TCC0506-025MLEA 74.79 USD
TCC0506-025ML TCC0506-500ML
Cyclooctene ≥95.0%
Cyclooctene

CAS Number: 931-88-4 MDL Number: MFCD00001753 Molecular Formula: C8H14 Molecular Weight: 110.20 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 144 Flash Point (°C): 24 Specific Gravity (20/20): 0.85

Formula: C₈H₁₄
Boiling Pt: 144 °C
Melting Pt: –16 °C
Density: 0.85 (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00001753
CAS Number: 931-88-4
EINECS: 213-245-5
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82825246c524f89a6d7dffdd53d829689a36da06&vid=64b431a5db2f05ae61af5218f5e508a12df34ef8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR