Crotonic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid
C0416-25G
C0416-100G
TCC0416-025GEA
70.25
USD
TCC0416-025G
TCC0416-100G
Crotonic acid ≥98.0% (by GC, titration analysis)
Crotonic acid
CAS Number: 107-93-7
MDL Number: MFCD00002701
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 71
Flash Point (°C): 89
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 185 °C Melting Pt: 71 °C Density: 1.02 g/cm³ (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00002701 CAS Number: 107-93-7 EINECS: 203-533-9 UN: 2823 ADR: 8,III Merck Index: 13,02625 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...