Trimethyl citrate ≥98.0%

Supplier: TCI America

Synonyms: Citric acid trimethyl ester

C0368-25G C0368-5G
TCC0368-025GEA 106 USD
TCC0368-025G TCC0368-5G
Trimethyl citrate ≥98.0%
Trimethyl citrate

CAS Number: 1587-20-8
MDL Number: MFCD00025889
Molecular Formula: C9H14O7
Molecular Weight: 234.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 176
Melting point (°C): 79


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Citric Acid Trimethyl Ester

Formula: C₉H₁₄O₇
MW: 234.21 g/mol
Boiling Pt: 176 °C
Melting Pt: 79 °C
MDL Number: MFCD00025889
CAS Number: 1587-20-8
EINECS: 216-449-2

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR