2-Chloro-p-xylene ≥98.0%
Supplier: TCI America
CAS Number: 95-72-7
MDL Number: MFCD00000564
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 57
Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2,5-Dimethylchlorobenzene
Formula:
C₈H₉Cl Boiling Pt: 186 °C Melting Pt: 1.5 °C Density: 1.06 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00000564 CAS Number: 95-72-7 EINECS: 202-444-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...