1-Chloropropane ≥99.0%

Supplier: TCI America
Danger

C0266-25ML C0266-500ML
TCC0266-025MLEA 23.86 USD
TCC0266-025ML TCC0266-500ML
1-Chloropropane ≥99.0%
1-Chloropropane

CAS Number: 540-54-5
MDL Number: MFCD00000995
Molecular Formula: C3H7Cl
Molecular Weight: 78.54
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 46
Melting point (°C): -123
Flash Point (°C): -18
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Propyl Chloride

Formula: C₃H₇Cl
Boiling Pt: 46 °C
Melting Pt: –123 °C
Density: 0.89 (20 °C)
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000995
CAS Number: 540-54-5
EINECS: 208-749-7
UN: 1278
ADR: 3,II
Merck Index: 12,08032
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c7cc02518aa050be259d251ff4c8695727f8af9&vid=13117ccf0d1e0a728caa4bfcd9a58cf15615688c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR