(1-Chloroethyl)benzene ≥97.0%
Supplier: TCI America
CAS Number: 672-65-1
MDL Number: MFCD00018870
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 44
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 1-Phenylethyl Chloride
Formula:
C₆H₅CHClCH₃ |
MDL Number:
MFCD00018870 CAS Number: 672-65-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...