1-Chlorododecane ≥97.0%

Supplier: TCI America
Warning

Synonyms: Lauryl chloride, Dodecylchloride

C0163-500ML C0163-25ML
TCC0163-500MLEA 66.45 USD
TCC0163-500ML TCC0163-025ML
1-Chlorododecane ≥97.0%
1-Chlorododecane

CAS Number: 112-52-7
MDL Number: MFCD00013688
Molecular Formula: C12H25Cl
Molecular Weight: 204.78
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 132
Freezing point (°C): -9
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Dodecyl Chloride, Lauryl Chloride

Formula: C₁₂H₂₅Cl
MW: 204.78 g/mol
Boiling Pt: 260 °C
Melting Pt: –9 °C
Density: 0.87 (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8024b629a18030884bc8f716034ee400f0676ddc&vid=01fd71f8766a7a260f3cdb26d5df7b84956acb04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR