4-Chloro-m-cresol ≥99.0%

Supplier: TCI America
Danger

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

C0150-25G C0150-500G
TCC0150-025GEA 42 USD
TCC0150-025G TCC0150-500G
4-Chloro-m-cresol ≥99.0%
4-Chloro-m-cresol

CAS Number: 59-50-7
MDL Number: MFCD00002323
Molecular Formula: C7H7ClO
Molecular Weight: 142.58
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 66
Flash Point (°C): 118


Other Product Names / Synonyms: 4-Chloro-3-methylphenol

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 235 °C
Melting Pt: 66 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II
Merck Index: 13,02152
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21af2b9b19228f70767d409afc5327827cc15218&vid=21af2b9b19228f70767d409afc5327827cc15218&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR