1-Bromo-3-methoxypropane ≥98.0%
Supplier: TCI America
B3499-5G
B3499-25G
TCB3499-5GEA
21
USD
TCB3499-5G
TCB3499-25G
1-Bromo-3-methoxypropane ≥98.0%
1-Bromo-3-methoxypropane
CAS Number: 36865-41-5
MDL Number: MFCD02258473
Molecular Formula: C4H9BrO
Molecular Weight: 153.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Specific Gravity (20/20): 1.37
Other Product Names / Synonyms: 3-Bromopropyl Methyl Ether
Formula:
C₄H₉BrO Boiling Pt: 132 °C Density: 1.37 (20 °C) |
MDL Number:
MFCD02258473 CAS Number: 36865-41-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...