2-Bromo-4-fluorobenzaldehyde ≥98.0%
Supplier: TCI America
B3498-25G
B3498-5G
TCB3498-25GEA
90
USD
TCB3498-25G
TCB3498-5G
2-Bromo-4-fluorobenzaldehyde ≥98.0%
2-Bromo-4-fluorobenzaldehyde
CAS Number: 59142-68-6
MDL Number: MFCD00672923
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64
Formula:
C₇H₄BrFO MW: 203.01 g/mol Melting Pt: 64 °C |
MDL Number:
MFCD00672923 CAS Number: 59142-68-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...