1-Bromoisoquinoline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3445-5G B3445-1G
TCB3445-5GEA 604.7 USD
TCB3445-5G TCB3445-1G
1-Bromoisoquinoline ≥98.0% (by titrimetric analysis)
1-Bromoisoquinoline

CAS Number: 1532-71-4
MDL Number: MFCD00234478
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 44

Formula: C₉H₆BrN
Melting Pt: 44 °C
MDL Number: MFCD00234478
CAS Number: 1532-71-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f92407f080517465e0c4cd7f68bdb36c154d90&vid=29f92407f080517465e0c4cd7f68bdb36c154d90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR