1-Butyl-3-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-Butyl-3-methylpyridinium Chloride
B3425-1G
B3425-5G
TCB3425-1GEA
102
USD
TCB3425-1G
TCB3425-5G
1-Butyl-3-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)
1-Butyl-3-methylpyridinium chloride
CAS Number: 125652-55-3
MDL Number: MFCD08458942
Molecular Formula: C10H16ClN
Molecular Weight: 185.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₁₀H₁₆ClN MW: 185.7 g/mol |
MDL Number:
MFCD08458942 CAS Number: 125652-55-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...