4-Bromoanthranilic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 20776-50-5
MDL Number: MFCD03618454
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2-Amino-4-bromobenzoic Acid
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD03618454 CAS Number: 20776-50-5 EINECS: 244-025-7 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...