3-Bromo-5-chlorobenzonitrile ≥98.0%

Supplier: TCI America

B3118-1G B3118-5G
TCB3118-1GEA 41.8 USD
TCB3118-1G TCB3118-5G
3-Bromo-5-chlorobenzonitrile ≥98.0%
3-Bromo-5-chlorobenzonitrile

CAS Number: 304854-55-5
MDL Number: MFCD06200840
Molecular Formula: C7H3BrClN
Molecular Weight: 216.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 144
Melting point (°C): 74

Formula: C₇H₃BrClN
Boiling Pt: 144 °C
Melting Pt: 74 °C
MDL Number: MFCD06200840
CAS Number: 304854-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=908a91176898e1bd9a9a01f984eaa4d694edb55b&vid=908a91176898e1bd9a9a01f984eaa4d694edb55b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR