5-Bromo-2-chlorobenzonitrile ≥98.0%
Supplier: TCI America
B3116-25G
B3116-5G
TCB3116-25GEA
519
USD
TCB3116-25G
TCB3116-5G
5-Bromo-2-chlorobenzonitrile ≥98.0%
5-Bromo-2-chlorobenzonitrile
CAS Number: 57381-44-9
MDL Number: MFCD00672948
Molecular Formula: C7H3BrClN
Molecular Weight: 216.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133
Formula:
C₇H₃BrClN Melting Pt: 133 °C |
MDL Number:
MFCD00672948 CAS Number: 57381-44-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...