4-Bromo-p-terphenyl ≥97.0%

Supplier: TCI America

B2155-1G B2155-5G
TCB2155-1GEA 68 USD
TCB2155-1G TCB2155-5G
4-Bromo-p-terphenyl ≥97.0%
4-Bromo-p-terphenyl

CAS Number: 1762-84-1
MDL Number: MFCD02685117
Molecular Formula: C18H13Br
Molecular Weight: 309.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 232

Formula: C₁₈H₁₃Br
MW: 309.1998 g/mol
Melting Pt: 232 °C
MDL Number: MFCD02685117
CAS Number: 1762-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8b055fcaac2518e47e2791b754fd8d03778beec&vid=f8b055fcaac2518e47e2791b754fd8d03778beec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR