4-tert-Butyl-2-nitrophenol ≥98.0%

Supplier: TCI America

Synonyms: 4-(tert-Butyl)-2-nitrophenol

B1812-5G B1812-25G
TCB1812-5GEA 64.9 USD
TCB1812-5G TCB1812-025G
4-tert-Butyl-2-nitrophenol ≥98.0%
4-tert-Butyl-2-nitrophenol

CAS Number: 3279-07-0
MDL Number: MFCD00191888
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 109
Specific Gravity (20/20): 1.13

Formula: C₁₀H₁₃NO₃
MW: 195.2151 g/mol
Density: 1.13 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00191888
CAS Number: 3279-07-0
EINECS: 221-914-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403e15f61f76bd2c7280ecc991b491b8a5bc71ba&vid=403e15f61f76bd2c7280ecc991b491b8a5bc71ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR