(R)-(+)-2-Bromopropionic acid ≥98.0%

Supplier: TCI America

B1756-5G B1756-25G
TCB1756-005GEA 145 USD
TCB1756-005G TCB1756-25G
(R)-(+)-2-Bromopropionic acid ≥98.0%
(R)-(+)-2-Bromopropionic acid

CAS Number: 10009-70-8
MDL Number: MFCD00145205
Molecular Formula: C3H5BrO2
Molecular Weight: 152.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 100
Specific Gravity (20/20): 1.70
Specific rotation [a]20/D: 27 deg (neat)

Formula: H₃CBrCHCOOH
Boiling Pt: 104 °C
Melting Pt: 25 °C
Density: 1.70 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145205
CAS Number: 10009-70-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4e1b6ed5f73b8454defde9acccd775c48b190e&vid=2f4e1b6ed5f73b8454defde9acccd775c48b190e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR