2-(Butylamino)ethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Butylethanolamine

B1023-25ML B1023-500ML
TCB1023-025MLEA 22.39 USD
TCB1023-025ML TCB1023-500ML
2-(Butylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(Butylamino)ethanol

CAS Number: 111-75-1
MDL Number: MFCD00020570
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 77
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: N-Butylethanolamine

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199 °C
Density: 0.89 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00020570
CAS Number: 111-75-1
EINECS: 203-904-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d611884b4e7db5ea8dc38b690490d0726e59296c&vid=d611884b4e7db5ea8dc38b690490d0726e59296c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR