1-Bromo-4-methylpentane ≥98.0%

Supplier: TCI America

Synonyms: 4-Methylpentyl bromide

B1017-5ML B1017-25ML
TCB1017-005MLEA 170.9 USD
TCB1017-005ML TCB1017-25ML
1-Bromo-4-methylpentane ≥98.0%
1-Bromo-4-methylpentane

CAS Number: 626-88-0
MDL Number: MFCD00013544
Molecular Formula: C6H13Br
Molecular Weight: 165.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 57
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 4-Methylpentyl Bromide

Formula: C₆H₁₃Br
Boiling Pt: 148 °C
Melting Pt: -71 °C
Density: 1.17 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00013544
CAS Number: 626-88-0
EINECS: 210-968-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=055e68190bc488cf9d4b667f61f788cfc944cb26&vid=055e68190bc488cf9d4b667f61f788cfc944cb26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR