1-Bromotridecane ≥98.0%

Supplier: TCI America

Synonyms: Tridecyl bromide

B0935-5ML B0935-25ML
TCB0935-5MLEA 127.2 USD
TCB0935-5ML TCB0935-025ML
1-Bromotridecane ≥98.0%
1-Bromotridecane

CAS Number: 765-09-3
MDL Number: MFCD00000227
Molecular Formula: C13H27Br
Molecular Weight: 263.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Freezing point (°C): 5
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Tridecyl Bromide

Formula: BrH₂C(CH₂)₁₁CH₃
MW: 263.26 g/mol
Boiling Pt: 150 °C
Melting Pt: 6 °C
Density: 1.03 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000227
CAS Number: 765-09-3
EINECS: 212-138-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c41469044b0a954f93405e762ec47606e421f05&vid=f544e04685c023a25d0e9add243286daf9f753d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR