Bromoacetyl chloride ≥85.0%

Supplier: TCI America

B0900-25G B0900-500G
TCB0900-025GEA 71.24 USD
TCB0900-025G TCB0900-500G
Bromoacetyl chloride ≥85.0%
Bromoacetyl chloride

CAS Number: 22118-09-8
MDL Number: MFCD00000724
Molecular Formula: C2H2BrClO
Molecular Weight: 157.39
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Specific Gravity (20/20): 1.88
Storage Temperature: 0-10°C

Formula: BrCH₂COOCl
MW: 157.39 g/mol
Boiling Pt: 128 °C
Density: 1.88 (20 °C)
MDL Number: MFCD00000724
CAS Number: 22118-09-8
EINECS: 244-790-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=553e1ec9ea6ba296d96133bd44372e2a2c98d30e&vid=553e1ec9ea6ba296d96133bd44372e2a2c98d30e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR