Isobutyl butyrate ≥98.0%

Supplier: TCI America

Synonyms: Butyric acid Isobutyl ester, 2-Methyl-1-propylbutyrate

B0761-25ML B0761-500ML
TCB0761-025MLEA 17 USD
TCB0761-025ML TCB0761-500ML
Isobutyl butyrate ≥98.0%
Isobutyl butyrate

CAS Number: 539-90-2
MDL Number: MFCD00027134
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 49
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Butyric Acid Isobutyl Ester

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 157 °C
Density: 0.86 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00027134
CAS Number: 539-90-2
EINECS: 208-729-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1dee5510dfc5aca5d5c8a3f82f993ab4d7216e&vid=7e1dee5510dfc5aca5d5c8a3f82f993ab4d7216e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR