4-tert-Butyltoluene ≥95.0%
Supplier: TCI America
Synonyms:
1-(tert-Butyl)-4-methylbenzene
B0744-25ML
B0744-500ML
TCB0744-025MLEA
23
USD
TCB0744-025ML
TCB0744-500ML
4-tert-Butyltoluene ≥95.0%
4-tert-Butyltoluene
CAS Number: 98-51-1
MDL Number: MFCD00008837
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 61
Specific Gravity (20/20): 0.86
Formula:
C₁₁H₁₆ MW: 148.25 g/mol Boiling Pt: 193 °C Melting Pt: -62.5 °C Density: 0.86 (20 °C) Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008837 CAS Number: 98-51-1 EINECS: 202-675-9 UN: 2667 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...