6-tert-Butyl-o-cresol ≥99.0%

Supplier: TCI America

Synonyms: 2-(tert-Butyl)-6-methylphenol, 2-tert-Butyl-6-methylphenol

B0728-500G B0728-25G
TCB0728-500GEA 198 USD
TCB0728-500G TCB0728-025G
6-tert-Butyl-o-cresol ≥99.0%
6-tert-Butyl-o-cresol

CAS Number: 2219-82-1
MDL Number: MFCD00002239
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 107
Freezing point (°C): 28


Other Product Names / Synonyms: 2-tert-Butyl-6-methylphenol

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 30 - 32 °C
Density: 0.967 g/cm³ (25 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002239
CAS Number: 2219-82-1
EINECS: 218-734-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a8e3aca2917e7627c6b7f29cd70543ade20691&vid=63a8e3aca2917e7627c6b7f29cd70543ade20691&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR