6-tert-Butyl-m-cresol ≥97.0%

Supplier: TCI America

Synonyms: 2-tert-Butyl-5-methylphenol, 5-Methyl-2-tert-butylphenol

B0718-500G B0718-25G
TCB0718-500GEA 126 USD
TCB0718-500G TCB0718-025G
6-tert-Butyl-m-cresol ≥97.0%
6-tert-Butyl-m-cresol

CAS Number: 88-60-8
MDL Number: MFCD00002308
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 244
Flash Point (°C): 118
Freezing point (°C): 21


Other Product Names / Synonyms: 2-tert-Butyl-5-methylphenol, 5-Methyl-2-tert-butylphenol

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 244 °C
Melting Pt: 20…23 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00002308
CAS Number: 88-60-8
EINECS: 201-842-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5a7e37242163383e8f4d4ef0c039a1cc783dcf&vid=6d5a7e37242163383e8f4d4ef0c039a1cc783dcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR