tert-Butylbenzene ≥98.0%

Supplier: TCI America

B0715-25ML B0715-500ML
TCB0715-025MLEA 26.75 USD
TCB0715-025ML TCB0715-500ML
tert-Butylbenzene ≥98.0%
tert-Butylbenzene

CAS Number: 98-06-6
MDL Number: MFCD00008816
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 60
Specific Gravity (20/20): 0.87

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 168 °C
Melting Pt: -58 °C
Density: 0.87 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008816
CAS Number: 98-06-6
EINECS: 202-632-4
UN: 2709
ADR: 3,III
Merck Index: 13,01550
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d333cf9d1c7b4ca0a425c1ff964d9d4e9c8e489&vid=b44b3b4eb06714aba0da6d0971c4ddf8495936a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR