2-Bromoisobutyric acid 98,0

Supplier: TCI America

Synonyms: 2-Bromo-2-methylpropanoic acid, α-Bromoisobutyric acid, 2-Bromo-2-methylpropionic acid

B0605-25G B0605-500G B0605-100G
TCB0605-025GEA 34 USD
TCB0605-025G TCB0605-500G TCB0605-100G
2-Bromoisobutyric acid 98,0
2-Bromoisobutyric acid

CAS Number: 2052-01-9
MDL Number: MFCD00004168
Molecular Formula: C4H7BrO2
Molecular Weight: 167.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 46
Flash Point (°C): 110

Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 200 °C
Melting Pt: 48
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004168
CAS Number: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95a23b0c8daf88e442a920300e5d5154bd6c2657&vid=95a23b0c8daf88e442a920300e5d5154bd6c2657&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR