Benzyltriethylammonium iodide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
N-Benzyl-N,N-diethylethanaminium iodide
B0446-25G
B0446-5G
TCB0446-025GEA
128.7
USD
TCB0446-025G
TCB0446-5G
Benzyltriethylammonium iodide ≥98.0% (by titrimetric analysis)
Benzyltriethylammonium iodide
CAS Number: 5400-94-2
MDL Number: MFCD00043178
Molecular Formula: C13H22IN
Molecular Weight: 319.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 171
Formula:
C₁₃H₂₂IN MW: 319.23 g/mol Melting Pt: 171 °C |
MDL Number:
MFCD00043178 CAS Number: 5400-94-2 EINECS: 226-446-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...