Benzoyl bromide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0104-25G B0104-5G
TCB0104-025GEA 172.4 USD
TCB0104-025G TCB0104-5G
Benzoyl bromide ≥98.0% (by GC, titration analysis)
Benzoyl bromide

CAS Number: 618-32-6
MDL Number: MFCD00000112
Molecular Formula: C7H5BrO
Molecular Weight: 185.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 90
Freezing point (°C): -24
Specific Gravity (20/20): 1.55

Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 219 °C
Melting Pt: 0 °C
Density: 1.55 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000112
CAS Number: 618-32-6
EINECS: 210-544-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcafaa8db28bdfc47a42098af19978178621921a&vid=fcafaa8db28bdfc47a42098af19978178621921a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR