Benz[a]anthracene ≥98.0%

Supplier: TCI America
Danger

B0017-5G B0017-1G B0017-25G
TCB0017-005GEA 429 USD
TCB0017-005G TCB0017-001G TCB0017-25G
Benz[a]anthracene ≥98.0%
Benz[a]anthracene

CAS Number: 56-55-3
MDL Number: MFCD00003599
Molecular Formula: C18H12
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 438
Melting point (°C): 161


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Benzanthrene, Tetraphene

Formula: C₁₈H₁₂
Boiling Pt: 438 °C
Melting Pt: 161 °C
Density: 1.245 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003599
CAS Number: 56-55-3
EINECS: 200-280-6
UN: 3077
ADR: 9,III
Merck Index: 12,01089
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=259bb3deeeac4b6166e2f5b5d5698942fdabf509&vid=9423af22bc73b496cb2cd82305d392f1c96d035a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR