4-Amino-3-fluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A2164-1G A2164-5G
TCA2164-1GEA 72.25 USD
TCA2164-1G TCA2164-5G
4-Amino-3-fluorophenol ≥98.0% (by GC, titration analysis)
4-Amino-3-fluorophenol

CAS Number: 399-95-1
MDL Number: MFCD00077452
Molecular Formula: C6H6FNO
Molecular Weight: 127.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 139


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-4-hydroxyaniline

Formula: C₆H₆FNO
Melting Pt: 139 °C
MDL Number: MFCD00077452
CAS Number: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bec6d37e19f8e9a5ed27cfd254c11581787b54&vid=a9bec6d37e19f8e9a5ed27cfd254c11581787b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR