Allyltrimethoxysilane ≥97.0%

Supplier: TCI America

Synonyms: Trimethoxyallylsilane

A1504-25ML A1504-5ML
TCA1504-025MLEA 132 USD
TCA1504-025ML TCA1504-005ML
Allyltrimethoxysilane ≥97.0%
Allyltrimethoxysilane

CAS Number: 2551-83-9
MDL Number: MFCD00053865
Molecular Formula: C6H14O3Si
Molecular Weight: 162.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 31
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trimethoxyallylsilane

Formula: H₂CCHCH₂Si(OCH₃)₃
MW: 162.26 g/mol
Boiling Pt: 136 °C
Density: 0.97 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00053865
CAS Number: 2551-83-9
EINECS: 219-855-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95a6b43f87109bd99233754ac44771959aea7d3a&vid=95a6b43f87109bd99233754ac44771959aea7d3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR